2-Bromo-6-methylbenzaldehyde 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-2-formyltoluene, 6-Bromo-o-tolualdehyde
OR43527-1G OR43527-250MG OR43527-5G
APOSOR43527-1GEA 244 EUR
APOSOR43527-1G APOSOR43527-250MG APOSOR43527-5G
2-Bromo-6-methylbenzaldehyde 96%
2-Bromo-6-methylbenzaldehyde
Formula: C₈H₇BrO
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD18392003
CAS Number: 176504-70-4