2'-Methoxy-3'-methylacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 3-Acetyl-2-methoxytoluene, 2-Acetyl-6-methylanisole, 1-(2-Methoxy-3-methylphenyl)ethan-1-one
OR43670-1G OR43670-250MG OR43670-5G
APOSOR43670-1GEA 174 EUR
APOSOR43670-1G APOSOR43670-250MG APOSOR43670-5G
2'-Methoxy-3'-methylacetophenone
2'-Methoxy-3'-methylacetophenone
Formula: C₁₀H₁₂O₂
Boiling Pt: 240 °C
Density: 1.061 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD11044853
CAS Number: 6342-75-2