2-Amino-5-bromo-4-(tert-butyl)-1,3-thiazole hydrobromide 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-4-(tert-butyl)-1,3-thiazol-2-amine hydrobromide
OR4382-1G
OR4382-5G
OR4382-10G
APOSOR4382-1GEA
87
EUR
APOSOR4382-1G
APOSOR4382-5G
APOSOR4382-10G
2-Amino-5-bromo-4-(tert-butyl)-1,3-thiazole hydrobromide 98%
2-Amino-5-bromo-4-(tert-butyl)-1,3-thiazole hydrobromide
Formula:
C₇H₁₁BrN₂S·BrH Boiling Pt: 110…112 °C ( 0.3 mm Hg) Melting Pt: 235 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052240 CAS Number: 175136-77-3 |