5-(Bromoacetyl)benzo[b]thiophene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 5-(Bromoacetyl)-1-benzothiophene, 1-(Benzo[b]thiophen-5-yl)-2-bromoethan-1-one
OR4407-1G OR4407-250MG
APOSOR4407-1GEA 169 EUR
APOSOR4407-1G APOSOR4407-250MG
5-(Bromoacetyl)benzo[b]thiophene 97%
5-(Bromoacetyl)benzo[b]thiophene
Formula: C₁₀H₇BrOS
MW: 255.131 g/mol
Boiling Pt: 118…121 °C (20 mm Hg)
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD07368508
CAS Number: 1131-87-9
UN: 3261
ADR: 8 III