2-Chloro-3-nitroimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
OR450073-1G OR450073-250MG
APOSOR450073-1GEA 864 EUR
APOSOR450073-1G APOSOR450073-250MG
2-Chloro-3-nitroimidazo[1,2-a]pyridine
2-Chloro-3-nitroimidazo[1,2-a]pyridine
Formula: C₇H₄ClN₃O₂
MW: 197.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07343825
CAS Number: 4926-52-7