Ethyl 7-hydroxytetrazolo[1,5-a]pyrimidine-6-carboxylate
Supplier: Apollo Scientific
OR450108-1G OR450108-250MG
APOSOR450108-1GEA 429 EUR
APOSOR450108-1G APOSOR450108-250MG
Ethyl 7-hydroxytetrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl 7-hydroxytetrazolo[1,5-a]pyrimidine-6-carboxylate
Formula: C₇H₇N₅O₃
MW: 209.162 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32264085
CAS Number: 1778637-73-2