2-sulphanylacetohydrazide
Supplier: Apollo Scientific
|
Danger
|
OR450115-1G
OR450115-250MG
APOSOR450115-1GEA
244
EUR
APOSOR450115-1G
APOSOR450115-250MG
2-sulphanylacetohydrazide
2-sulphanylacetohydrazide
Formula:
C₂H₆N₂OS MW: 106.147 g/mol Melting Pt: 50…53 °C Storage Temperature: Freezer |
MDL Number:
MFCD18263132 CAS Number: 760-30-5 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...