2-Iodo-6-nitrophenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Hydroxy-3-iodonitrobenzene
OR45114-1G OR45114-250MG OR45114-5G
APOSOR45114-1GEA 186 EUR
APOSOR45114-1G APOSOR45114-250MG APOSOR45114-5G
2-Iodo-6-nitrophenol 97%
2-Iodo-6-nitrophenol
Formula: C₆H₄INO₃
MW: 265.005 g/mol
Melting Pt: 109…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00859244
CAS Number: 13073-26-2