8-(Bromomethyl)-1,4-dioxaspiro[4.5]decane
Supplier: Apollo Scientific
Warning
OR451150-1G OR451150-250MG
APOSOR451150-1GEA 734 EUR
APOSOR451150-1G APOSOR451150-250MG
8-(Bromomethyl)-1,4-dioxaspiro[4.5]decane
8-(Bromomethyl)-1,4-dioxaspiro[4.5]decane
Formula: C₉H₁₅BrO₂
MW: 235.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847773
CAS Number: 74286-87-6