1,4-Dioxaspiro[4.5]decane-8-methanol
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Dioxaspiro[4.5]decan-8-ylmethanol
OR451159-1G OR451159-250MG OR451159-5G OR451159-10G
APOSOR451159-1GEA 42.2 EUR
APOSOR451159-1G APOSOR451159-250MG APOSOR451159-5G APOSOR451159-10G
1,4-Dioxaspiro[4.5]decane-8-methanol
1,4-Dioxaspiro[4.5]decane-8-methanol
Formula: C₉H₁₆O₃
MW: 172.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779441
CAS Number: 17159-82-9