1-Phenylcyclobutanamine
Supplier: Apollo Scientific
OR451172-100MG OR451172-1G OR451172-250MG
APOSOR451172-100MGEA 256 EUR
APOSOR451172-100MG APOSOR451172-1G APOSOR451172-250MG
1-Phenylcyclobutanamine
1-Phenylcyclobutanamine
Formula: C₁₀H₁₃N
MW: 147.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08444192
CAS Number: 17380-77-7