4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 4,6-Dichloro-5-azaindazole
OR45126-100MG OR45126-1G OR45126-250MG
APOSOR45126-100MGEA 62.5 EUR
APOSOR45126-100MG APOSOR45126-1G APOSOR45126-250MG
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₃Cl₂N₃
MW: 188.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257114
CAS Number: 1256794-28-1