Pyrazolo[1,5-a]pyrimidin-7-amine
Supplier: Apollo Scientific
Warning
OR451452-1G OR451452-5G
APOSOR451452-1GEA 197 EUR
APOSOR451452-1G APOSOR451452-5G
Pyrazolo[1,5-a]pyrimidin-7-amine
Pyrazolo[1,5-a]pyrimidin-7-amine
Formula: C₆H₆N₄
MW: 134.139 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01693048
CAS Number: 1194-63-4