N-Phenylcyanamide

Supplier: Apollo Scientific
Warning

OR45177-1G OR45177-250MG
APOSOR45177-1GEA 676 EUR
APOSOR45177-1G APOSOR45177-250MG
N-Phenylcyanamide
N-Phenylcyanamide
Formula: C₇H₆N₂
MW: 118.136 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00235035
CAS Number: 622-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR