3-(N-Boc-N-methylamino)benzoic acid 97%
Supplier: Apollo Scientific
OR452048-1G OR452048-500MG OR452048-5G
APOSOR452048-1GEA 236 EUR
APOSOR452048-1G APOSOR452048-500MG APOSOR452048-5G
3-(N-Boc-N-methylamino)benzoic acid 97%
3-(N-Boc-N-methylamino)benzoic acid
Formula: C₁₃H₁₇NO₄
MW: 251.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848820
CAS Number: 168162-30-9