4-[1-(Boc-amino)ethyl]benzoic acid 97%
Supplier: Apollo Scientific
Synonyms:
4-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid
OR452050-1G
APOSOR452050-1GEA
130
EUR
APOSOR452050-1G
4-[1-(Boc-amino)ethyl]benzoic acid 97%
4-[1-(Boc-amino)ethyl]benzoic acid
Formula:
C₁₄H₁₉NO₄ MW: 265.305 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11505982 CAS Number: 895577-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...