5-(Cyclopropyl)thiophene-2-carboxaldehyde
Supplier: Apollo Scientific
OR45208-1G OR45208-250MG
APOSOR45208-1GEA 654 EUR
APOSOR45208-1G APOSOR45208-250MG
5-(Cyclopropyl)thiophene-2-carboxaldehyde
5-(Cyclopropyl)thiophene-2-carboxaldehyde
Formula: C₈H₈OS
MW: 152.214 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06802412
CAS Number: 29481-26-3