3,3-Dimethyl-morpholine hydrochloride 97%
Supplier: Apollo Scientific
OR452081-1G OR452081-250MG OR452081-500MG
APOSOR452081-1GEA 654 EUR
APOSOR452081-1G APOSOR452081-250MG APOSOR452081-500MG
3,3-Dimethyl-morpholine hydrochloride 97%
3,3-Dimethyl-morpholine hydrochloride
Formula: C₆H₁₃NO·ClH
MW: 151.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052542
CAS Number: 59229-64-0