Fmoc-(S)-3-amino-2-hydroxypropionic acid 97%
Supplier: Apollo Scientific
OR452098-1G OR452098-250MG OR452098-500MG
APOSOR452098-1GEA 783 EUR
APOSOR452098-1G APOSOR452098-250MG APOSOR452098-500MG
Fmoc-(S)-3-amino-2-hydroxypropionic acid 97%
Fmoc-(S)-3-amino-2-hydroxypropionic acid
Formula: C₁₈H₁₇NO₅
MW: 327.331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18252739
CAS Number: 172721-23-2