2-(Cyclopropyl)-5-(hydroxymethyl)thiophene
Supplier: Apollo Scientific
OR45210-1G OR45210-250MG
APOSOR45210-1GEA 766 EUR
APOSOR45210-1G APOSOR45210-250MG
2-(Cyclopropyl)-5-(hydroxymethyl)thiophene
2-(Cyclopropyl)-5-(hydroxymethyl)thiophene
Formula: C₈H₁₀OS
MW: 154.229 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28664217
CAS Number: 1456863-44-7