3-(4-(Tert-butyl)phenyl)propan-1-ol 97%
Supplier: Apollo Scientific
OR452136-1G OR452136-500MG OR452136-5G
APOSOR452136-1GEA 261 EUR
APOSOR452136-1G APOSOR452136-500MG APOSOR452136-5G
3-(4-(Tert-butyl)phenyl)propan-1-ol 97%
3-(4-(Tert-butyl)phenyl)propan-1-ol
Formula: C₁₃H₂₀O
MW: 192.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464127
CAS Number: 78574-08-0