Cbz-DL-Prolinol 97%
Supplier: Apollo Scientific
OR452153-1G OR452153-5G
APOSOR452153-1GEA 184 EUR
APOSOR452153-1G APOSOR452153-5G
Cbz-DL-Prolinol 97%
Cbz-DL-Prolinol
Formula: C₁₃H₁₇NO₃
MW: 235.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953280
CAS Number: 86954-05-4