N1,N1,3-Trimethyl-1,2-butanediamine 97%
Supplier: Apollo Scientific
OR452191-1G OR452191-250MG OR452191-500MG
APOSOR452191-1GEA 1050 EUR
APOSOR452191-1G APOSOR452191-250MG APOSOR452191-500MG
N1,N1,3-Trimethyl-1,2-butanediamine 97%
N1,N1,3-Trimethyl-1,2-butanediamine
Formula: C₇H₁₈N₂
MW: 130.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11155327
CAS Number: 773045-76-4