N-Boc-[(R)-1-(aminomethyl)-2-methylpropyl]amine 97%
Supplier: Apollo Scientific
OR452203-1G OR452203-250MG OR452203-500MG
APOSOR452203-1GEA 870 EUR
APOSOR452203-1G APOSOR452203-250MG APOSOR452203-500MG
N-Boc-[(R)-1-(aminomethyl)-2-methylpropyl]amine 97%
N-Boc-[(R)-1-(aminomethyl)-2-methylpropyl]amine
Formula: C₁₀H₂₂N₂O₂
MW: 202.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08726032
CAS Number: 400652-50-8