(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: (1R,2R)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine
OR460002-250MG OR460002-1G OR460002-5G
APOSOR460002-250MGEA 79.9 EUR
APOSOR460002-250MG APOSOR460002-1G APOSOR460002-5G
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95+%
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine
Formula: C₁₀H₂₂N₂
MW: 170.295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411208
CAS Number: 53152-69-5