5-Bromobenzene-1,3-diamine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromophenylene-1,3-diamine, 1-Bromo-3,5-diaminobenzene
OR46062-1G OR46062-250MG OR46062-5G
APOSOR46062-1GEA 91.5 EUR
APOSOR46062-1G APOSOR46062-250MG APOSOR46062-5G
5-Bromobenzene-1,3-diamine
5-Bromobenzene-1,3-diamine
Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Melting Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09036306
CAS Number: 33786-90-2
UN: 2811
ADR: 6.1,III