2,4-Dibromobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
OR46091-1G OR46091-5G
APOSOR46091-1GEA 21.9 EUR
APOSOR46091-1G APOSOR46091-5G
2,4-Dibromobenzaldehyde 98%
2,4-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
MW: 263.914 g/mol
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD13185489
CAS Number: 5629-98-1