Methyl 1-[4-(chlorosulphonyl)phenyl]cyclobutanecarboxylate, Technical Grade
Supplier: Apollo Scientific
Danger
OR46261-1G OR46261-250MG
APOSOR46261-1GEA 575 EUR
APOSOR46261-1G APOSOR46261-250MG
Methyl 1-[4-(chlorosulphonyl)phenyl]cyclobutanecarboxylate, Technical Grade
Methyl 1-[4-(chlorosulphonyl)phenyl]cyclobutanecarboxylate
Formula: C₁₂H₁₃ClO₄S
MW: 288.747 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32219476
CAS Number: 2407349-26-0
UN: 3261
ADR: 8,III