3-Bromoisoxazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-1,2-oxazole
OR46500-1G OR46500-250MG OR46500-5G
APOSOR46500-1GEA 114 EUR
APOSOR46500-1G APOSOR46500-250MG APOSOR46500-5G
3-Bromoisoxazole
3-Bromoisoxazole
Formula: C₃H₂BrNO
MW: 147.958 g/mol
Boiling Pt: 182…183 °C
Density: 1.812 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD07778379
CAS Number: 111454-71-8
UN: 1993
ADR: 3 III