Methyl 2-Bromo-5-hydroxy-4-methylbenzoate
Supplier: Apollo Scientific
OR46691-100MG OR46691-250MG OR46691-500MG
APOSOR46691-100MGEA 236 EUR
APOSOR46691-100MG APOSOR46691-250MG APOSOR46691-500MG
Methyl 2-Bromo-5-hydroxy-4-methylbenzoate
Methyl 2-Bromo-5-hydroxy-4-methylbenzoate
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976442
CAS Number: 87808-27-3