3-Chloro-6-methyl-4-nitropyridazine
Supplier: Apollo Scientific
Warning
OR46740-1G OR46740-250MG OR46740-5G
APOSOR46740-1GEA 522 EUR
APOSOR46740-1G APOSOR46740-250MG APOSOR46740-5G
3-Chloro-6-methyl-4-nitropyridazine
3-Chloro-6-methyl-4-nitropyridazine
Formula: C₅H₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18377816
CAS Number: 771-78-8