3-Bromo-1-tert-butyl-pyrazolo[3,4-d]pyrimidin-4-ol
Supplier: Apollo Scientific
Warning
OR46760-1G OR46760-250MG
APOSOR46760-1GEA 208 EUR
APOSOR46760-1G APOSOR46760-250MG
3-Bromo-1-tert-butyl-pyrazolo[3,4-d]pyrimidin-4-ol
3-Bromo-1-tert-butyl-pyrazolo[3,4-d]pyrimidin-4-ol
Formula: C₉H₁₁BrN₄O
MW: 271.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30487814
CAS Number: 2044713-81-5