cis-1-Methylhexahydropyrrolo[3,4-b]pyrrole dihydrochloride
Supplier: Apollo Scientific
OR470016-1G OR470016-5G
APOSOR470016-1GEA 194 EUR
APOSOR470016-1G APOSOR470016-5G
cis-1-Methylhexahydropyrrolo[3,4-b]pyrrole dihydrochloride
cis-1-Methylhexahydropyrrolo[3,4-b]pyrrole dihydrochloride
Formula: C₇H₁₄N₂·2ClH
MW: 199.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21364418
CAS Number: 1234805-59-4