3-Bromo-1-methylpiperidine hydrobromide
Supplier: Apollo Scientific
OR470080-5G OR470080-1G
APOSOR470080-5GEA 1220 EUR
APOSOR470080-5G APOSOR470080-1G
3-Bromo-1-methylpiperidine hydrobromide
3-Bromo-1-methylpiperidine hydrobromide
Formula: C₆H₁₂BrN·BrH
MW: 258.982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375234
CAS Number: 13617-02-2