3-(2-Methoxyphenoxy)propylamine
Supplier: Apollo Scientific
OR470093-1G OR470093-5G
APOSOR470093-1GEA 256 EUR
APOSOR470093-1G APOSOR470093-5G
3-(2-Methoxyphenoxy)propylamine
3-(2-Methoxyphenoxy)propylamine
Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05053657
CAS Number: 3245-88-3