(S)-1-Phenyl-2-propyn-1-ol
Supplier: Apollo Scientific
OR470118-1G
OR470118-250MG
APOSOR470118-1GEA
499
EUR
APOSOR470118-1G
APOSOR470118-250MG
(S)-1-Phenyl-2-propyn-1-ol
(S)-1-Phenyl-2-propyn-1-ol
Formula:
C₉H₈O MW: 132.159 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210084 CAS Number: 64599-56-0 |