(R)-1-Phenyl-2-propyn-1-ol
Supplier: Apollo Scientific
OR470119-1G OR470119-5G
APOSOR470119-1GEA 306 EUR
APOSOR470119-1G APOSOR470119-5G
(R)-1-Phenyl-2-propyn-1-ol
(R)-1-Phenyl-2-propyn-1-ol
Formula: C₉H₈O
MW: 132.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210083
CAS Number: 61317-73-5