Methyl 2-Methyl-3-(methylsulphonyl)benzoate
Supplier: Apollo Scientific
Warning
OR470139-5G OR470139-1G
APOSOR470139-5GEA 712 EUR
APOSOR470139-5G APOSOR470139-1G
Methyl 2-Methyl-3-(methylsulphonyl)benzoate
Methyl 2-Methyl-3-(methylsulphonyl)benzoate
Formula: C₁₀H₁₂O₄S
MW: 228.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18071000
CAS Number: 1256633-15-4