2-(2-Pyridyl)-2-propylamine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Pyridin-2-yl)propan-2-amine
OR470173-1G OR470173-5G OR470173-250MG
APOSOR470173-1GEA 137 EUR
APOSOR470173-1G APOSOR470173-5G APOSOR470173-250MG
2-(2-Pyridyl)-2-propylamine 97%
2-(2-Pyridyl)-2-propylamine
Formula: C₈H₁₂N₂
Density: 0.98 g/cm³ (25 °C)
Flash Pt: 79.44 °C
Storage Temperature: Ambient
MDL Number: MFCD08729302
CAS Number: 52568-28-2