Methyl-4-iodo-2-methoxybenzoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate
OR4703-25G
OR4703-5G
OR4703-100G
APOSOR4703-25GEA
168
EUR
APOSOR4703-25G
APOSOR4703-5G
APOSOR4703-100G
Methyl-4-iodo-2-methoxybenzoate
Methyl 4-iodo-2-methoxybenzoate
Formula:
C₉H₉IO₃ Melting Pt: 44…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052925 CAS Number: 148490-97-5 |