4,5-Dimethoxy-2-(methylsulphonamido)benzoic acid
Supplier: Apollo Scientific
OR470316-1G OR470316-5G
APOSOR470316-1GEA 245 EUR
APOSOR470316-1G APOSOR470316-5G
4,5-Dimethoxy-2-(methylsulphonamido)benzoic acid
4,5-Dimethoxy-2-(methylsulphonamido)benzoic acid
Formula: C₁₀H₁₃NO₆S
MW: 275.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09045487
CAS Number: 926239-35-2