3-Methyl-2-(methylsulphonamido)benzoic acid
Supplier: Apollo Scientific
OR470318-1G OR470318-5G
APOSOR470318-1GEA 245 EUR
APOSOR470318-1G APOSOR470318-5G
3-Methyl-2-(methylsulphonamido)benzoic acid
3-Methyl-2-(methylsulphonamido)benzoic acid
Formula: C₉H₁₁NO₄S
MW: 229.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09045450
CAS Number: 926206-21-5