1-[8-(3,4-Dimethoxyphenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
Supplier: Apollo Scientific
OR470335-1G OR470335-5G
APOSOR470335-1GEA 489 EUR
APOSOR470335-1G APOSOR470335-5G
1-[8-(3,4-Dimethoxyphenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
1-[8-(3,4-Dimethoxyphenyl)-1,4-dioxaspiro[4.5]dec-8-yl]methanamine
Formula: C₁₇H₂₅NO₄
MW: 307.385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410584
CAS Number: 887978-91-8