3-(1-Cbz-4-piperidyl)-2-(Boc-amino)propanoic acid
Supplier: Apollo Scientific
Synonyms: Boc-DL-Ala(β-1-Cbz-Piperidine-4-yl)-OH, 3-(1-((Benzyloxy)carbonyl)piperidin-4-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid
OR470342-1G OR470342-5G
APOSOR470342-1GEA 794 EUR
APOSOR470342-1G APOSOR470342-5G
3-(1-Cbz-4-piperidyl)-2-(Boc-amino)propanoic acid
3-(1-Cbz-4-piperidyl)-2-(Boc-amino)propanoic acid
Formula: C₂₁H₃₀N₂O₆
MW: 406.473 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179070
CAS Number: 195877-90-8