4-(Aminomethyl)-4-(3-chlorophenyl)cyclohexanol
Supplier: Apollo Scientific
OR470361-1G OR470361-5G
APOSOR470361-1GEA 499 EUR
APOSOR470361-1G APOSOR470361-5G
4-(Aminomethyl)-4-(3-chlorophenyl)cyclohexanol
4-(Aminomethyl)-4-(3-chlorophenyl)cyclohexanol
Formula: C₁₃H₁₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD06410569
CAS Number: 887978-40-7