[6-(2,4-Dimethoxyphenyl)-2-pyridyl]methanol
Supplier: Apollo Scientific
OR470375-1G OR470375-5G
APOSOR470375-1GEA 601 EUR
APOSOR470375-1G APOSOR470375-5G
[6-(2,4-Dimethoxyphenyl)-2-pyridyl]methanol
[6-(2,4-Dimethoxyphenyl)-2-pyridyl]methanol
Formula: C₁₄H₁₅NO₃
MW: 245.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410133
CAS Number: 887981-31-9