Methyl 3-(1-Methyl-5-imidazolyl)benzoate
Supplier: Apollo Scientific
OR470422-1G OR470422-5G
APOSOR470422-1GEA 306 EUR
APOSOR470422-1G APOSOR470422-5G
Methyl 3-(1-Methyl-5-imidazolyl)benzoate
Methyl 3-(1-Methyl-5-imidazolyl)benzoate
Formula: C₁₂H₁₂N₂O₂
MW: 216.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20491413
CAS Number: 1346597-46-3