4-(Methylamino)piperidine dihydrochloride
Supplier: Apollo Scientific
Synonyms: N-Methylpiperidin-4-amine dihydrochloride
OR470425-25G OR470425-5G
APOSOR470425-25GEA 916 EUR
APOSOR470425-25G APOSOR470425-5G
4-(Methylamino)piperidine dihydrochloride
4-(Methylamino)piperidine dihydrochloride
Formula: C₆H₁₄N₂·2ClH
MW: 187.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039797
CAS Number: 1220039-56-4