Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate
Supplier: Apollo Scientific
OR470469-25G OR470469-5G
APOSOR470469-25GEA 287 EUR
APOSOR470469-25G APOSOR470469-5G
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate
Formula: C₁₅H₃₀O₅Si
MW: 318.481 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16251279
CAS Number: 91424-39-4