(1S,6R)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
Supplier: Apollo Scientific
OR470480-1G OR470480-250MG
APOSOR470480-1GEA 2860 EUR
APOSOR470480-1G APOSOR470480-250MG
(1S,6R)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
(1S,6R)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
Formula: C₁₄H₁₈N₂O₂
MW: 246.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22205851
CAS Number: 1293941-04-4